Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AHP4
Entrez Gene ID
820883
Gene Name
HPT phosphotransmitter 4
Gene Aliases
AT3G16360, HPT phosphotransmitter 4, T2O4.20
Chromosome Location
Chr.3: 5554197 - 5555789 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202983.2 NP_001189912.1 2 - 3 331 140
NM_112507.1 NP_566544.1 1 - 2 169 140
GenBank mRNA AB041766.1 - 1 - 2 169 140

View Full Product Details