Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPPalpha
Entrez Gene ID
820896
Gene Name
mitochondrial processing peptidase alpha subunit
Gene Aliases
AT3G16480, T2O4.13, mitochondrial processing peptidase alpha subunit
Chromosome Location
Chr.3: 5599710 - 5603027 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112519.4 NP_566548.1 7 - 8 1247 63
GenBank mRNA AY142643.1 - 7 - 8 1051 63

View Full Product Details