The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112550.5 | NP_188299.1 | 1 - 2 | 398 | 87 |
GenBank mRNA | AF003096.1 | - | 1 - 2 | 224 | 87 |
AY035100.1 | - | 1 - 2 | 185 | 87 | |
AY059917.1 | - | 1 - 2 | 195 | 87 | |
AY087488.1 | - | 1 - 2 | 216 | 87 | |
AY142562.1 | - | 1 - 2 | 155 | 87 |