Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPR2
Entrez Gene ID
820936
Gene Name
TOPLESS-related 2
Gene Aliases
AT3G16830, TOPLESS-related 2
Chromosome Location
Chr.3: 5731519 - 5737854 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112557.4 NP_188306.2 10 - 11 1461 81

View Full Product Details