Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRPB9A
Entrez Gene ID
820954
Gene Name
RNA polymerases M/15 Kd subunit
Gene Aliases
AT3G16980, NRPD9A, NRPE9A
Chromosome Location
Chr.3: 5794392 - 5795827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112574.3 NP_188323.1 3 - 4 399 76
GenBank mRNA BT012109.1 - 3 - 4 328 76
BT012363.1 - 3 - 4 265 76

View Full Product Details