Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POK1
Entrez Gene ID
820999
Gene Name
phragmoplast orienting kinesin 1
Gene Aliases
AT3G17360, phragmoplast orienting kinesin 1
Chromosome Location
Chr.3: 5935880 - 5946540 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338286.1 NP_001326050.1 26 - 27 5525 104
NM_001338287.1 NP_001326049.1 26 - 27 5484 104
NM_112614.4 NP_188362.2 26 - 27 5501 104
GenBank mRNA DQ399529.1 - 26 - 27 5273 104

View Full Product Details