Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPK1
Entrez Gene ID
821006
Gene Name
glyoxysomal protein kinase 1
Gene Aliases
AT3G17420, glyoxysomal protein kinase 1
Chromosome Location
Chr.3: 5959174 - 5962289 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112621.6 NP_188368.2 4 - 5 1337 91
GenBank mRNA AY065430.1 - 4 - 5 1202 91
AY096380.1 - 3 - 4 966 91

View Full Product Details