Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CIPK1
- Entrez Gene ID
-
821016
- Gene Name
- CBL-interacting protein kinase 1
- Gene Aliases
-
AT3G17510, CBL-interacting protein kinase 1, SNF1-RELATED PROTEIN KINASE 3.16, SnRK3.16
- Chromosome Location
-
Chr.3: 5988924 - 5993001 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112631.3 |
NP_566580.1 |
1 - 2 |
557 |
71 |
View Full Product Details