Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BSH
Entrez Gene ID
821025
Gene Name
transcription regulatory protein SNF5, putative (BSH)
Gene Aliases
AT3G17590, BUSHY, BUSHY GROWTH, CHE1
Chromosome Location
Chr.3: 6017166 - 6019724 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202989.1 NP_001189918.1 9 - 10 701 98
NM_001338294.1 NP_001326663.1 8 - 9 714 98
NM_112639.4 NP_566581.1 9 - 10 753 98
GenBank mRNA AK117162.1 - 9 - 10 695 98
BT024620.1 - 9 - 10 620 98

View Full Product Details