Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHX19
Entrez Gene ID
821030
Gene Name
cation/H+ exchanger 19
Gene Aliases
AT3G17630, ATCHX19, cation/H+ exchanger 19
Chromosome Location
Chr.3: 6028688 - 6031986 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338295.1 NP_001319577.1 2 - 3 551 61
NM_112644.3 NP_188390.2 1 - 2 401 61
GenBank mRNA AY926475.1 - 1 - 2 203 61

View Full Product Details