Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNBT1
Entrez Gene ID
821037
Gene Name
cyclic nucleotide-binding transporter 1
Gene Aliases
AT3G17700, ATCNGC20, CNGC20, CYCLIC NUCLEOTIDE-GATED CHANNEL 20, cyclic nucleotide gated channel 20, cyclic nucleotide-binding transporter 1
Chromosome Location
Chr.3: 6048814 - 6052675 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338307.1 NP_001326341.1 7 - 8 1722 107
NM_112651.5 NP_566585.1 7 - 8 1820 107
GenBank mRNA AF148541.1 - 7 - 8 1699 107

View Full Product Details