Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP17
Entrez Gene ID
821047
Gene Name
purple acid phosphatase 17
Gene Aliases
AT3G17790, ATACP5, ATPAP17, purple acid phosphatase 17
Chromosome Location
Chr.3: 6089381 - 6091231 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112660.6 NP_566587.1 1 - 2 596 85
GenBank mRNA AJ133747.1 - 1 - 2 320 85
AK228106.1 - 1 - 2 260 85
AY818189.1 - 1 - 2 198 85
BT003135.1 - 1 - 2 198 85

View Full Product Details