Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TDX
Entrez Gene ID
821056
Gene Name
tetraticopeptide domain-containing thioredoxin
Gene Aliases
ARABIDOPSIS THALIANA HSP70-INTERACTING PROTEIN 2, AT3G17870, AT3G17880, ATHIP2, ATTDX, HIP, HSC-70 INTERACTING PROTEIN, tetraticopeptide domain-containing thioredoxin
Chromosome Location
Chr.3: 6123341 - 6126281 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084706.1 NP_001078175.1 4 - 5 477 100
NM_112669.4 NP_188415.2 4 - 5 566 100
GenBank mRNA AK175494.1 - 4 - 5 438 100
AK227340.1 - 4 - 5 461 100
AY064251.1 - 4 - 5 446 100

View Full Product Details