Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOC64-III
Entrez Gene ID
819660
Gene Name
translocon at the outer membrane of chloroplasts 64-III
Gene Aliases
AT3G17960, AT3G17970, atToc64-III, translocon at the outer membrane of chloroplasts 64-III
Chromosome Location
Chr.3: 6147906 - 6152082 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338317.1 NP_001325422.1 11 - 12 1862 82
NM_112678.4 NP_188424.2 12 - 13 1806 82

View Full Product Details