Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUB29
Entrez Gene ID
821402
Gene Name
plant U-box 29
Gene Aliases
ARABIDOPSIS THALIANA PLANT U-BOX 29, AT3G18710, ATPUB29, plant U-box 29
Chromosome Location
Chr.3: 6434082 - 6436046 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112757.3 NP_188501.1 1 - 1 1759 88

View Full Product Details