Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPAT5
Entrez Gene ID
821418
Gene Name
lysophosphatidyl acyltransferase 5
Gene Aliases
AT3G18850, lysophosphatidyl acyltransferase 5
Chromosome Location
Chr.3: 6499332 - 6501432 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035647.3 NP_001030724.2 3 - 4 956 76
NM_001084714.1 NP_001078183.1 3 - 4 1037 76
NM_001084715.1 NP_001078184.1 4 - 5 800 76
NM_001338361.1 NP_001326795.1 4 - 5 315 76
NM_112771.5 NP_188515.1 3 - 4 942 76
NM_202606.2 NP_974335.1 2 - 3 1151 76
GenBank mRNA AY099849.1 - 3 - 4 849 76
BT008462.1 - 2 - 3 681 76

View Full Product Details