Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PARC6
Entrez Gene ID
821452
Gene Name
plastid division protein
Gene Aliases
A. THALIANA CHLOROPLAST DIVISION SITE POSITIONING 1, ARC6H, AT3G19180, ATCDP1, CDP1, CHLOROPLAST DIVISION SITE POSITIONING 1, paralog of ARC6
Chromosome Location
Chr.3: 6632637 - 6637292 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035648.1 NP_001030725.1 1 - 2 433 68
NM_112805.5 NP_188549.2 1 - 2 472 68

View Full Product Details