The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035648.1 | NP_001030725.1 | 5 - 6 | 918 | 92 |
NM_112805.5 | NP_188549.2 | 5 - 6 | 957 | 92 |