Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOH2
Entrez Gene ID
821460
Gene Name
LAG1 homologue 2
Gene Aliases
AT3G19260, LAG1 HOMOLOG 2, LAG1 homologue 2, LONGEVITY ASSURANCE GENE1 HOMOLOG 2
Chromosome Location
Chr.3: 6669065 - 6671638 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112813.2 NP_188557.1 5 - 6 1246 136
GenBank mRNA AK118029.1 - 5 - 6 1098 136
AY084980.1 - 5 - 6 1128 136

View Full Product Details