Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHD1
Entrez Gene ID
821483
Gene Name
sodium:hydrogen antiporter 1
Gene Aliases
ARABIDOPSIS THALIANA NA/H ANTIPORTER 1, AT3G19490, ATNHD1, sodium:hydrogen antiporter 1
Chromosome Location
Chr.3: 6754615 - 6758285 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112836.4 NP_566638.1 8 - 9 1479 87
GenBank mRNA AY084922.1 - 8 - 9 1455 87

View Full Product Details