Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT6
Entrez Gene ID
821541
Gene Name
protein arginine methyltransferase 6
Gene Aliases
ARABIDOPSIS THALIANA PROTEIN ARGININE METHYLTRANSFERASE 6, AT3G20020, ATPRMT6, protein arginine methyltransferase 6
Chromosome Location
Chr.3: 6983702 - 6988101 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084722.1 NP_001078191.1 10 - 11 1231 75
NM_001338447.1 NP_001326463.1 11 - 12 1299 75
NM_112893.4 NP_188637.2 11 - 12 1297 75
GenBank mRNA BT029008.1 - 11 - 12 1143 75

View Full Product Details