Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT6
Entrez Gene ID
821541
Gene Name
protein arginine methyltransferase 6
Gene Aliases
ARABIDOPSIS THALIANA PROTEIN ARGININE METHYLTRANSFERASE 6, AT3G20020, ATPRMT6, protein arginine methyltransferase 6
Chromosome Location
Chr.3: 6983702 - 6988101 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084722.1 NP_001078191.1 7 - 8 847 121
NM_001338447.1 NP_001326463.1 8 - 9 915 121
NM_112893.4 NP_188637.2 8 - 9 913 121
GenBank mRNA BT029008.1 - 8 - 9 759 121

View Full Product Details