Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EHD1
Entrez Gene ID
821573
Gene Name
EPS15 homology domain 1
Gene Aliases
AT3G20290, ATEHD1, EPS15 homology domain 1
Chromosome Location
Chr.3: 7074764 - 7079014 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035654.2 NP_001030731.1 7 - 8 587 79
NM_001338474.1 NP_001327849.1 8 - 9 665 79
NM_112920.4 NP_566657.1 8 - 9 732 79
GenBank mRNA AK317308.1 - 7 - 8 653 79
AY034911.1 - 8 - 9 589 79
BT004414.1 - 7 - 8 459 79

View Full Product Details