Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SVL3
- Entrez Gene ID
-
821598
- Gene Name
- SHV3-like 3
- Gene Aliases
-
AT3G20520, GDPDL5, Glycerophosphodiester phosphodiesterase (GDPD) like 5, SHV3-like 3
- Chromosome Location
-
Chr.3: 7162766 - 7165864 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112944.3 |
NP_188688.1 |
3 - 4 |
538 |
136 |
View Full Product Details