Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHT7
Entrez Gene ID
829671
Gene Name
LYS/HIS transporter 7
Gene Aliases
AT4G35180, LYS/HIS transporter 7, T12J5.50, T12J5_50
Chromosome Location
Chr.4: 16737835 - 16740601 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342328.1 NP_001328042.1 3 - 4 1035 97
NM_119684.4 NP_567977.2 3 - 4 1035 97

View Full Product Details