Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NRPB1
- Entrez Gene ID
-
829734
- Gene Name
- RNA polymerase II large subunit
- Gene Aliases
-
AT4G35800, F4B14.70, F4B14_70, RNA POLYMERASE II LARGE SUBUNIT, RNA polymerase II large subunit, RNA_POL_II_LS, RNA_POL_II_LSRNA_POL_II_LS, RPB1
- Chromosome Location
-
Chr.4: 16960558 - 16968128 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119746.4 |
NP_195305.2 |
13 - 14 |
5780 |
79 |
View Full Product Details