Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARO3
Entrez Gene ID
829759
Gene Name
armadillo repeat only 3
Gene Aliases
AT4G36030, T19K4.160, T19K4_160, armadillo repeat only 3
Chromosome Location
Chr.4: 17044875 - 17047315 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119770.3 NP_195327.1 1 - 1 2125 98
GenBank mRNA AK117432.1 - 1 - 1 1955 98
BT004921.1 - 1 - 1 1906 98

View Full Product Details