Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH3F1
Entrez Gene ID
829782
Gene Name
aldehyde dehydrogenase 3F1
Gene Aliases
AT4G36250, F23E13.140, F23E13_140, aldehyde dehydrogenase 3F1
Chromosome Location
Chr.4: 17150740 - 17153663 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119793.5 NP_195348.2 3 - 4 867 66
GenBank mRNA BT030380.1 - 3 - 4 578 66

View Full Product Details