Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROT3
Entrez Gene ID
829790
Gene Name
Cytochrome P450 superfamily protein
Gene Aliases
AP22.10, AP22_10, AT4G36380, CYP90C1, ROTUNDIFOLIA 3
Chromosome Location
Chr.4: 17187528 - 17192507 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342408.1 NP_001328464.1 1 - 2 549 107
NM_119801.4 NP_568002.1 1 - 2 611 107
GenBank mRNA BT029220.1 - 1 - 2 306 107

View Full Product Details