Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROT3
Entrez Gene ID
829790
Gene Name
Cytochrome P450 superfamily protein
Gene Aliases
AP22.10, AP22_10, AT4G36380, CYP90C1, ROTUNDIFOLIA 3
Chromosome Location
Chr.4: 17187528 - 17192507 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342408.1 NP_001328464.1 6 - 7 1637 123
NM_119801.4 NP_568002.1 7 - 8 1627 123
GenBank mRNA BT029220.1 - 7 - 8 1322 123

View Full Product Details