Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP81D4
- Entrez Gene ID
-
829888
- Gene Name
- cytochrome P450, family 81, subfamily D, polypeptide 4
- Gene Aliases
-
''cytochrome P450, AT4G37330, F6G17.5, cytochrome P450, family 81, polypeptide 4, polypeptide 4'', subfamily D
- Chromosome Location
-
Chr.4: 17562317 - 17564763 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119897.3 |
NP_195450.1 |
2 - 3 |
1068 |
109 |
GenBank mRNA |
BT030464.1 |
- |
2 - 3 |
874 |
109 |
View Full Product Details