Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LBD39
Entrez Gene ID
829909
Gene Name
LOB domain-containing protein 39
Gene Aliases
AT4G37540, F19F18.30, F19F18_30, LOB domain-containing protein 39
Chromosome Location
Chr.4: 17639393 - 17640925 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119918.3 NP_195470.1 1 - 2 601 68
GenBank mRNA AB473874.1 - 1 - 2 309 68
AY072144.1 - 1 - 2 326 68
AY122976.1 - 1 - 2 185 68

View Full Product Details