Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HKL3
Entrez Gene ID
829940
Gene Name
hexokinase-like 3
Gene Aliases
AT4G37840, T28I19.120, T28I19_120, hexokinase-like 3
Chromosome Location
Chr.4: 17790080 - 17792198 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119945.2 NP_195497.1 5 - 6 934 70

View Full Product Details