Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK26
Entrez Gene ID
829980
Gene Name
calcium-dependent protein kinase 26
Gene Aliases
ARABIDOPSIS THALIANA CALCIUM-DEPENDENT PROTEIN KINASE 26, AT4G38230, ATCPK26, F20D10.350, F20D10_350, calcium-dependent protein kinase 26
Chromosome Location
Chr.4: 17928677 - 17931511 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204020.2 NP_001190949.1 7 - 8 1513 112
NM_001204021.1 NP_001190950.1 8 - 9 1369 112
NM_001342486.1 NP_001328909.1 7 - 8 1364 112
NM_119985.2 NP_195536.2 7 - 8 1227 112
GenBank mRNA BT029228.1 - 5 - 6 681 112

View Full Product Details