Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAUT3
Entrez Gene ID
829984
Gene Name
galacturonosyltransferase 3
Gene Aliases
AT4G38270, F22I13.40, F22I13_40, galacturonosyltransferase 3
Chromosome Location
Chr.4: 17938372 - 17941717 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204023.1 NP_001190952.1 4 - 5 229 67
NM_119989.5 NP_195540.2 4 - 5 263 67
GenBank mRNA KJ138714.1 - 4 - 5 202 67

View Full Product Details