Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSL03
Entrez Gene ID
817755
Gene Name
glucan synthase-like 3
Gene Aliases
AT2G31960, ATGSL03, ATGSL3, F22D22.29, F22D22_29, glucan synthase-like 3
Chromosome Location
Chr.2: 13588522 - 13600430 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202724.2 NP_001189653.1 10 - 11 1822 91
NM_001336361.1 NP_001318331.1 11 - 12 1534 91
NM_001336362.1 NP_001323730.1 10 - 11 1411 91
NM_001336363.1 NP_001323731.1 11 - 12 1459 91
NM_179847.3 NP_850178.2 10 - 11 1462 91

View Full Product Details