Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arpc2b
Entrez Gene ID
817902
Gene Name
actin-related protein C2B
Gene Aliases
AT2G33385, actin-related protein C2B
Chromosome Location
Chr.2: 14147382 - 14149902 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202737.2 NP_001189666.1 2 - 3 261 85
NM_001336451.1 NP_001323477.1 2 - 3 261 85
NM_001336452.1 NP_001323478.1 2 - 3 259 85
NM_179877.1 NP_850208.1 2 - 3 128 85

View Full Product Details