Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE25
Entrez Gene ID
818050
Gene Name
NAD(P)-binding Rossmann-fold superfamily protein
Gene Aliases
AT2G34850, F19I3.8, F19I3_8, maternal effect embryo arrest 25
Chromosome Location
Chr.2: 14704507 - 14707425 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336531.1 NP_001324452.1 7 - 8 1184 83
NM_001336532.1 NP_001324451.1 8 - 9 1103 83
NM_001336533.1 NP_001324453.1 2 - 3 402 83
NM_179907.5 NP_850238.1 4 - 5 560 83
GenBank mRNA AY070487.1 - 4 - 5 571 83
AY091699.1 - 2 - 3 317 83

View Full Product Details