Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMZ3
Entrez Gene ID
818179
Gene Name
MMS ZWEI homologue 3
Gene Aliases
AT2G36060, F11F19.3, F11F19_3, MMS ZWEI homologue 3, UBIQUITIN E2 VARIANT 1C, UEV1C
Chromosome Location
Chr.2: 15142653 - 15144221 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084542.1 NP_001078011.1 1 - 2 215 58
NM_129165.4 NP_565834.1 1 - 2 254 58
NM_179928.2 NP_850259.1 1 - 2 215 58
GenBank mRNA AY062760.1 - 1 - 2 149 58
AY093382.1 - 1 - 2 31 58

View Full Product Details