Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAX1
Entrez Gene ID
818395
Gene Name
cation exchanger 1
Gene Aliases
AT2G38170, ATCAX1, F16M14.10, F16M14_10, HIGH AFFINITY CALCIUM ANTIPORTER CAX1, RARE COLD INDUCIBLE 4, RCI4, cation exchanger 1
Chromosome Location
Chr.2: 15989083 - 15993326 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129373.4 NP_181352.1 3 - 4 529 89
NM_179961.2 NP_850292.2 3 - 4 479 89
NM_201901.4 NP_973630.1 3 - 4 488 89
GenBank mRNA AF372938.1 - 3 - 4 476 89
AF461691.1 - 3 - 4 416 89
AK316894.1 - 3 - 4 480 89

View Full Product Details