Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAX1
Entrez Gene ID
818395
Gene Name
cation exchanger 1
Gene Aliases
AT2G38170, ATCAX1, F16M14.10, F16M14_10, HIGH AFFINITY CALCIUM ANTIPORTER CAX1, RARE COLD INDUCIBLE 4, RCI4, cation exchanger 1
Chromosome Location
Chr.2: 15989083 - 15993326 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_129373.4 NP_181352.1 8 - 9 1371 76
NM_179961.2 NP_850292.2 7 - 8 1425 76
NM_201901.4 NP_973630.1 8 - 9 1330 76
GenBank mRNA AF372938.1 - 8 - 9 1318 76
AK316894.1 - 8 - 9 1322 76

View Full Product Details