The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_100028.3 | NP_171653.2 | 4 - 5 | 734 | 86 |
GenBank mRNA | BT028973.1 | - | 3 - 4 | 413 | 86 |
DQ056443.1 | - | 3 - 4 | 413 | 86 |