Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GPAT4
- Entrez Gene ID
-
839297
- Gene Name
- glycerol-3-phosphate acyltransferase 4
- Gene Aliases
-
AT1G01610, ATGPAT4, F22L4.15, F22L4_15, GLYCEROL-3-PHOSPHATE ACYLTRANSFERASE 4, GLYCEROL-3-PHOSPHATE sn-2-ACYLTRANSFERASE 4, glycerol-3-phosphate acyltransferase 4
- Chromosome Location
-
Chr.1: 221642 - 224351 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100043.5 |
NP_171667.1 |
2 - 3 |
724 |
68 |
View Full Product Details