Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAD1
Entrez Gene ID
839232
Gene Name
GRAM domain family protein
Gene Aliases
AT1G02120, T7I23.28, T7I23_28, VASCULAR ASSOCIATED DEATH1
Chromosome Location
Chr.1: 395689 - 400001 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331325.1 NP_001321166.1 16 - 17 1739 116
NM_001331326.1 NP_001321165.1 16 - 17 1633 116
NM_100092.3 NP_171714.2 17 - 18 1638 116

View Full Product Details