Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAN1
Entrez Gene ID
839444
Gene Name
Glycosyl hydrolase superfamily protein
Gene Aliases
AT1G02310, T6A9.1, T6A9_1, endo-beta-mannanase 1
Chromosome Location
Chr.1: 458133 - 460831 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331331.1 NP_001321462.1 3 - 4 807 107
NM_100112.3 NP_171733.2 3 - 4 672 107
GenBank mRNA AY081352.1 - 3 - 4 672 107
BT002154.1 - 3 - 4 628 107

View Full Product Details