Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ABCB12
- Entrez Gene ID
-
839282
- Gene Name
- P-glycoprotein 12
- Gene Aliases
-
AT1G02530, ATP-binding cassette B12, P-glycoprotein 12, PGP12, T14P4.14, T14P4_14
- Chromosome Location
-
Chr.1: 529679 - 534677 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100134.4 |
NP_171754.1 |
7 - 8 |
3058 |
74 |
View Full Product Details