Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEA
Entrez Gene ID
839422
Gene Name
SET domain-containing protein
Gene Aliases
AT1G02580, EMB173, EMBRYO DEFECTIVE 173, FERTILIZATION INDEPENDENT SEED 1, FIS1, MEDEA, SDG5, SET DOMAIN-CONTAINING PROTEIN 5, T14P4.11, T14P4_11
Chromosome Location
Chr.1: 544733 - 549202 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331345.1 NP_001322965.1 12 - 13 1621 74
NM_100139.4 NP_563658.1 12 - 13 1621 74
GenBank mRNA AF060485.1 - 12 - 13 1571 74

View Full Product Details