Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RAB28
- Entrez Gene ID
-
839571
- Gene Name
- responsive to abscisic acid 28
- Gene Aliases
-
AT1G03120, ATRAB28, F10O3.5, F10O3_5, responsive to abscisic acid 28
- Chromosome Location
-
Chr.1: 752191 - 753140 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_100194.2 |
NP_171811.1 |
1 - 2 |
476 |
92 |
View Full Product Details