The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197972.1 | NP_001184901.1 | 2 - 3 | 466 | 76 |
NM_001331438.1 | NP_001318915.1 | 2 - 3 | 461 | 76 | |
NM_001331440.1 | NP_001323368.1 | 2 - 3 | 437 | 76 | |
NM_001331441.1 | NP_001323367.1 | 1 - 2 | 183 | 76 | |
NM_100256.3 | NP_171873.2 | 2 - 3 | 441 | 76 | |
GenBank mRNA | AK226183.1 | - | 2 - 3 | 443 | 76 |