Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRP39
Entrez Gene ID
839314
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT1G04080, F20D22.14
Chromosome Location
Chr.1: 1051312 - 1056827 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197976.1 NP_001184905.1 3 - 4 561 79
NM_001331460.1 NP_001322270.1 2 - 3 616 79
NM_100289.4 NP_563700.1 3 - 4 530 79
GenBank mRNA AY056321.1 - 3 - 4 495 79
AY087878.1 - 3 - 4 497 79
AY091187.1 - 2 - 3 347 79

View Full Product Details